missing translation for 'onlineSavingsMsg'
Få mere at vide

n-Tetratriacontane, SPEX CertiPrep™

Artikelnummer. 16339447 Shop alle SPEX Certiprep produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16339447 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16339447 Leverandør SPEX Certiprep Leverandørnr. S3485

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

Indeholder

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
n-Tetratriacontane14167-59-01000μg/mL

Certificeringer

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Overholdelse

GHS/OSHA Complaint

Tekniske data

Kemisk navn eller materiale n-Tetratriacontane
Emballage 2mL amber prescored glass ampule
CAS 14167-59-0, 75-09-2
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C34H70
MDL nummer MFCD00009412
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle GWVDBZWVFGFBCN-UHFFFAOYSA-N
SMIL CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
IUPAC navn tetratriacontane
Opløsningsmiddel eller Matrix MeCl
Molekylvægt (g/mol) 478.93
PubChem CID 26519
Mængde 1 mL
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • n-Tetratriacontane
Signal Word
  • Fare
Hazard Category
  • Akut toksicitet Kategori 4
  • Carcinogenicitet Kategori 2
  • Brandfarlig væske Kategori 2
Hazard Statement
  • H225-Meget brandfarlig væske og damp.
  • H302-Farlig ved indtagelse.
  • H351-Mistænkt for at fremkalde kræft.
Precautionary Statement
  • P210-Holdes væk fra varme, varme overflader, gnister, åben ild og andre antændelseskilder. Rygning forbudt.
  • P241-Anvend eksplosionssikkert elektrisk/ventilations-/lys-/ /udstyr.
  • P260-Indånd ikke pulver/røg/gas/tåge/damp/spray.
  • P303+P361+P353-VED KONTAKT MED HUDEN (eller håret): Alt tilsmudset tøj tages straks af. Skyl/brus huden med vand.
  • P405-Opbevares under lås.
  • P501b-Indholdet/beholderen bortskaffes i henhold til de lokale/regionale/nationale/internationale forskrifter.
Supplemental information
  • MIXTURE LIST-Contain: dichloromethane

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.