Få mere at vide
n-Tetradecyltrimethylammonium-d38 Bromide, TRC
Artikelnummer. 30467242
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30467242 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467242 Leverandør Toronto Research Chemicals Leverandørnr. TRCM8850422.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Tetradecyltrimethylammonium-D38 Bromide
CAS 95523-72-1
Molekylær formel C17D38BrN
Mængde 2.5 mg
Synonym 1-Tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29-aminium','N,N,N-tri(methyl-d3)-','bromide (9CI), Tetradecyltrimethylammonium bromide-d35, N,N,N-Tri(methyl-d3)-1-tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11
InChI formel InChI=1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2;
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H].[Br-]
IUPAC navn N,N,N-tris(methyl-d3)tetradecan-1-aminium-d29 bromide
Molekylvægt (g/mol) 374.63
Formel vægt 373.46