Få mere at vide
n-Tetradecyl-d29-trimethylammonium Bromide, CDN
Artikelnummer. 30326815
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326815 0.1 g 0.1g
30316467 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326815 Leverandør CDN Leverandørnr. CDND62500.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Tetradecyltrimethylammonium-D29 (tetradecyl-D29) Bromide
CAS 95523-73-2
Molekylær formel C17 D29 H9 N . Br
Mængde 0.1 g
Synonym 1-Tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29-aminium, N,N,N-trimethyl-, bromide (1:1) (ACI), 1-Tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29-aminium, N,N,N-trimethyl-, bromide (9 CI), N,N,N-Trimethyl-1-tetradecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-d29-aminium bromide, n-Tetradecyltrimethylammonium-d29 bromide, n-Tetradecyltrimethylammonium-D29 (tetradecyl-D29) bromide, (n-Tetradecyl-d29)trimethylammonium bromide
InChI formel InChI=1 S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1 H/q+1;/p-1/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2;
SMIL [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C)(C)C
IUPAC navn trimethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-nonacosadeuteriotetradecyl)azanium;bromide
Molekylvægt (g/mol) 365.573
Formel vægt 364.401 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity