missing translation for 'onlineSavingsMsg'
Få mere at vide

N-tert-Butyloxycarbonyl-L-aspartic Acid b-tert-Butyl Ester, TRC

Artikelnummer. 30335390
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30335390 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30335390 Leverandør Toronto Research Chemicals Leverandørnr. TRCB80920010G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-tert-Butyloxycarbonyl-L-aspartic Acid beta-tert-Butyl Ester
CAS 1676-90-0
Molekylær formel C13H23NO6
Mængde 10 g
Synonym tert-Butyloxycarbonyl-L-aspartic Acid 4-tert-Butyl Ester, L-N-Carboxyaspartic Acid N,4-di-tert-Butyl Ester, (S)-4-tert-Butoxy-2-[(tert-butoxycarbonyl)amino]-4-oxobutanoic Acid, 4-tert-Butyl (S)-2-tert-Butoxycarbonylaminosuccinate, BOC-L-Asp(Obut)-OH, Boc-L-Asp(OtBu)-OH, N-(tert-Butoxycarbonyl)-L-aspartic Acid beta-tert-Butyl Ester, N-Boc-L-asparaginic Acid gamma-(tert-Butyl) Ester, N-Boc-L-aspartic Acid beta-tert-Butyl Ester, N-tert-Butoxycarbonylaspartic Acid gamma-tert-Butyl Ester
InChI formel InChI=1S/C13H23NO6/c1-12(2,3)19-9(15)7-8(10(16)17)14-11(18)20-13(4,5)6/h8H,7H2,1-6H3,(H,14,18)(H,16,17)/t8-/m0/s1
SMIL CC(C)(C)OC(=O)C[C@H](NC(=O)OC(C)(C)C)C(=O)O
IUPAC navn (2S)-4-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Molekylvægt (g/mol) 289.32
Formel vægt 289.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.