missing translation for 'onlineSavingsMsg'
Få mere at vide

N-tert-Boc-L-aspartic Acid tert-Butyl Ester, TRC

Artikelnummer. 30340730
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340730 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30340730 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6213005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-tert-Boc-L-aspartic Acid tert-Butyl Ester
CAS 34582-32-6
Molekylær formel C13 H23 N O6
Mængde 5 g
InChI formel InChI=1S/C13H23NO6/c1-12(2,3)19-10(17)8(7-9(15)16)14-11(18)20-13(4,5)6/h8H,7H2,1-6H3,(H,14,18)(H,15,16)/t8-/m0/s1
SMIL CC(C)(C)OC(=O)N[C@@H](CC(=O)O)C(=O)OC(C)(C)C
IUPAC navn (3S)-4-[(2-methylpropan-2-yl)oxy]-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Molekylvægt (g/mol) 289.32
Formel vægt 289.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.