Få mere at vide
N-tert-Boc-L-alanine-D4, TRC
Artikelnummer. 30346240
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30346240 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346240 Leverandør Toronto Research Chemicals Leverandørnr. TRCB600402500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-tert-Boc-alanine-D4
CAS 714964-61-1
Molekylær formel C8 2H4 H11 N O4
Mængde 500 mg
Synonym L-Alanine-2,3,3,3-d4, N-[(1,1-dimethylethoxy)carbonyl]- (9CI, ACI), N-[(1,1-Dimethylethoxy)carbonyl]-L-alanine-2,3,3,3-d4 (ACI), (S)-2-((tert-Butoxycarbonyl)amino)propanoic-2,3,3,3-d4 acid, (2S)-2-(tert-Butoxycarbonylamino)propionic-2,3,3,3-d4 acid, (2S)-2-[(2-Methylpropan-2-yl)oxycarbonylamino]propanoic-2,3,3,3-d4 acid, N-BOC-L-alanine-2,3,3,3-d4, N-tert-Butoxycarbonyl-L-alanine-2,3,3,3-d4, N-(tert-Butoxycarbonyl)-L-alanine-2,3,3,3-D4, N-tert-Boc-L-alanine-2,3,3,3-d4, L-Alanine-2,3,3,3-d4-N-t-BOC, N-tert-Boc-L-alanine-D4
InChI formel InChI=1S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1/i1D3,5D
SMIL [2H]C([2H])([2H])[C@]([2H])(NC(=O)OC(C)(C)C)C(=O)O
IUPAC navn (2S)-2,3,3,3-tetradeuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Molekylvægt (g/mol) 193.23
Formel vægt 193.1252