Få mere at vide
N-tert-Boc-2-(4-hydroxy-2-methoxyphenoxy)ethylamine-d3, TRC
Artikelnummer. 30347518
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30347518 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347518 Leverandør Toronto Research Chemicals Leverandørnr. TRCB65650225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-tert-Boc-2-(4-hydroxy-2-methoxyphenoxy)ethylamine-d3
CAS 1189659-07-1
Molekylær formel C14H18D3NO5
Mængde 25 mg
Synonym [2-(4-Hydroxy-2-methoxyphenoxy-d3)ethyl]carbamic Acid 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C14H21NO5/c1-14(2,3)20-13(17)15-7-8-19-11-6-5-10(16)9-12(11)18-4/h5-6,9,16H,7-8H2,1-4H3,(H,15,17)/i4D3
SMIL [2H]C([2H])([2H])Oc1cc(O)ccc1OCCNC(=O)OC(C)(C)C
IUPAC navn tert-butyl N-[2-[4-hydroxy-2-(trideuteriomethoxy)phenoxy]ethyl]carbamate
Molekylvægt (g/mol) 286.34
Formel vægt 286.1608