Få mere at vide
N-t-Boc-valacyclovir-d4, TRC
Artikelnummer. 30331593
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30331593 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331593 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67350210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-t-Boc-valacyclovir-d4
CAS 1346617-11-5
Molekylær formel C18H24D4N6O6
Mængde 10 mg
Synonym N-T-Boc-Valacyclovir D4
InChI formel InChI=1S/C18H28N6O6/c1-10(2)11(21-17(27)30-18(3,4)5)15(26)29-7-6-28-9-24-8-20-12-13(24)22-16(19)23-14(12)25/h8,10-11H,6-7,9H2,1-5H3,(H,21,27)(H3,19,22,23,25)/t11-/m1/s1/i6D2,7D2
SMIL [2H]C([2H])(OCn1cnc2C(=O)NC(=Nc12)N)C([2H])([2H])OC(=O)[C@H](NC(=O)OC(C)(C)C)C(C)C
IUPAC navn [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-1,1,2,2-tetradeuterioethyl] (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
Molekylvægt (g/mol) 428.48
Formel vægt 428.2321