Få mere at vide
N-t-Boc-L-serine Allyl Ester, TRC
Artikelnummer. 30336822
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30336822 250 mg
30345856 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30336822 Leverandør Toronto Research Chemicals Leverandørnr. TRCB667375250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-t-Boc-L-serine Allyl Ester
CAS 143966-57-8
Molekylær formel C11 H19 N O5
Mængde 250 mg
Synonym L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 2-propen-1-yl ester, L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 2-propenyl ester (9CI), (S)-Allyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate, N-(tert-Butoxycarbonyl)serine allyl ester, N-(tert-Butoxycarbonyl)-L-serine allyl ester, N-t-Boc-L-serine allyl ester
InChI formel InChI=1S/C11H19NO5/c1-5-6-16-9(14)8(7-13)12-10(15)17-11(2,3)4/h5,8,13H,1,6-7H2,2-4H3,(H,12,15)/t8-/m0/s1
SMIL CC(C)(C)OC(=O)N[C@@H](CO)C(=O)OCC=C
IUPAC navn prop-2-enyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Molekylvægt (g/mol) 245.27
Formel vægt 245.1263