missing translation for 'onlineSavingsMsg'
Få mere at vide

N-t-Boc-L-serine Allyl Ester, TRC

Artikelnummer. 30346010
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30346010 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30346010 Leverandør Toronto Research Chemicals Leverandørnr. TRCB667375500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-t-Boc-L-serine Allyl Ester
CAS 143966-57-8
Molekylær formel C11 H19 N O5
Mængde 500 mg
Synonym L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 2-propen-1-yl ester, L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 2-propenyl ester (9CI), (S)-Allyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate, N-(tert-Butoxycarbonyl)serine allyl ester, N-(tert-Butoxycarbonyl)-L-serine allyl ester, N-t-Boc-L-serine allyl ester
InChI formel InChI=1S/C11H19NO5/c1-5-6-16-9(14)8(7-13)12-10(15)17-11(2,3)4/h5,8,13H,1,6-7H2,2-4H3,(H,12,15)/t8-/m0/s1
SMIL CC(C)(C)OC(=O)N[C@@H](CO)C(=O)OCC=C
IUPAC navn prop-2-enyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
Molekylvægt (g/mol) 245.27
Formel vægt 245.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.