missing translation for 'onlineSavingsMsg'
Learn More

N-Sulfamoylethyl Naratriptan Pyridinium Chloride, TRC

Product Code. 30334539
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30334539 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334539 Supplier Toronto Research Chemicals Supplier No. TRCB4861005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-[1,5-Bis-(2-methylsulfamoylethyl)-1H-indol-3-yl]-1-methylpyridinium Chloride
CAS 2144776-33-8
Molekylær formel C20H27ClN4O4S2
Mængde 5 mg
Synonym N-methyl-2-[3-(1-methylpyridin-1-ium-4-yl)-1-[2-(methylsulfamoyl)ethyl]indol-5-yl]ethanesulfonamide chloride
InChI formel InChI=1S/C20H27N4O4S2.ClH/c1-21-29(25,26)12-8-16-4-5-20-18(14-16)19(17-6-9-23(3)10-7-17)15-24(20)11-13-30(27,28)22-2;/h4-7,9-10,14-15,21-22H,8,11-13H2,1-3H3;1H/q+1;/p-1
SMIL O=S(CCC1=CC=C2C(C(C3=CC=[N+](C)C=C3)=CN2CCS(NC)(=O)=O)=C1)(NC)=O.[Cl-]
IUPAC navn N-methyl-2-[3-(1-methylpyridin-1-ium-4-yl)-1-[2-(methylsulfamoyl)ethyl]indol-5-yl]ethanesulfonamide;chloride
Molekylvægt (g/mol) 487.04
Formel vægt 486.1162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.