missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Succinimidyl N-methylcarbamate, 97%

Brand:  Alfa Aesar™ H63566.06

Additional Details : CAS Number : 18342-66-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15480319

  • 712.23 DKK / 5g


In stock, ready to ship: 1
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

18342-66-0
172.14
XMNGSPOWUCNRMO-UHFFFAOYSA-N
4381935
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
C6H8N2O4
MFCD00800292
2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
CHEBI:83949
CNC(=O)ON1C(=O)CCC1=O

Specifications

N-Succinimidyl N-methylcarbamate
5g
C6H8N2O4
1531863
Slightly soluble in water.
CNC(=O)ON1C(=O)CCC1=O
172.14
CHEBI:83949
Solid
Moisture sensitive
18342-66-0
MFCD00800292
2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
XMNGSPOWUCNRMO-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
4381935
172.14
97%
Videos
SDS Documents