missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate, 98+%

Product Code. 15431416
Change view
Click to view available options
Mængde:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
15431416 25 mg 25mg
15441416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431416 Supplier Thermo Scientific Alfa Aesar Supplier No. H54627.MB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 64987-85-5 | C16H18N2O6 | 334.328 g/mol

Denne Thermo Scientific Chemicals mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific Chemicals .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64987-85-5
Molekylær formel C16H18N2O6
Molekylvægt (g/mol) 334.328
MDL nummer MFCD00009634
InChI nøgle JJAHTWIKCUJRDK-UHFFFAOYSA-N
Synonym smcc, 2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate, trans-4-maleimidomethyl cyclohexanecarboxylic acid-nhs, unii-b357p1g1if, n-succinimidyl 4-maleimidomethyl cyclohexane-1-carboxylate, n-4-carboxycyclohexylmethyl maleimide n-hydroxysuccinimide ester, trans-2,5-dioxopyrrolidin-1-yl 4-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl methyl cyclohexanecarboxylate, n-hydroxysuccinimidyl 4-n-maleimidomethylcyclohexane-1-carboxylate, succinimidyl 4-n-maleimidomethyl cyclohexane-1-carboxylate
PubChem CID 125175
ChEBI CHEBI:63174
IUPAC navn (2,5-dioxopyrrolidin-1-yl) 4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexane-1-carboxylate
SMIL C1CC(CCC1CN2C(=O)C=CC2=O)C(=O)ON3C(=O)CCC3=O

Specifications

Kemisk navn eller materiale N-Succinimidyl 4-(maleimidomethyl)cyclohexanecarboxylate
Smeltepunkt 180°C to 182°C
Mængde 25 mg
Beilstein 1555271
Følsomhed Light and moisture sensitive
Formel vægt 334.33
Procent renhed ≥98%

RUO– Kun brug til forskning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.