Få mere at vide
N,S-Diacetyl-L-cysteine, Mikromol™
Artikelnummer. 17963586
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17963586 100 mg
17912716 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17963586 Leverandør Mikromol Leverandørnr. MM0024.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N,S-Diacetyl-L-cysteine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,S-Diacetyl-L-cysteine
CAS 18725-37-6
Analyt- eller komponentnavne N,S-Diacetyl-L-cysteine
Forsendelsestilstand Room Temperature
Molekylær formel C7 H11 N O4 S
Certificeringer/compliance ISO 17025
Synonym N,S-Diacetyl-L-cysteine, Acetylcysteine Imp. D (EP), Acetylcysteine Impurity D
InChI formel InChI=1S/C7H11NO4S/c1-4(9)8-6(7(11)12)3-13-5(2)10/h6H,3H2,1-2H3,(H,8,9)(H,11,12)/t6-/m0/s1
SMIL CC(=O)N[C@@H](CSC(=O)C)C(=O)O
IUPAC navn (2R)-2-acetamido-3-acetylsulfanylpropanoic acid
Molekylvægt (g/mol) 205.23
Mængde 100 mg
Formel vægt 205.0409 g/mol