Få mere at vide
N-[(S)-(4-Nitrophenoxy)phenoxy-2,3,4,5,6-d5-phosphinyl]-L-Alanine-d5 1-Methylethyl Ester, TRC
Artikelnummer. 30467382
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30467382 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467382 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49703250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(S)-(4-Nitrophenoxy)phenoxy-2,3,4,5,6-d5-phosphinyl]-L-Alanine-d5 1-Methylethyl Ester
CAS 1874196-88-9
Molekylær formel C18H16D5N2O7P
Mængde 50 mg
Synonym N-[(S)-(4-Nitrophenoxy)Phenoxy-2,3,4,5,6-D5-Phosphinyl]-L-Alanine-D5 1-Methylethyl Ester
InChI formel InChI=1S/C18H21N2O7P/c1-13(2)25-18(21)14(3)19-28(24,26-16-7-5-4-6-8-16)27-17-11-9-15(10-12-17)20(22)23/h4-14H,1-3H3,(H,19,24)/t14-,28?/m0/s1/i4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(OP(=O)(N[C@@H](C)C(=O)OC(C)C)Oc2ccc(cc2)[N+](=O)[O-])c([2H])c1[2H]
IUPAC navn propan-2-yl (2S)-2-[[(4-nitrophenoxy)-(2,3,4,5,6-pentadeuteriophenoxy)phosphoryl]amino]propanoate
Molekylvægt (g/mol) 413.37
Formel vægt 413.14