missing translation for 'onlineSavingsMsg'
Få mere at vide

N-[(S)-(4-Nitrophenoxy)phenoxy-2,3,4,5,6-d5-phosphinyl]-L-Alanine-d5 1-Methylethyl Ester, TRC

Artikelnummer. 30479815
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479815 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479815 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49703225MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-[(S)-(4-Nitrophenoxy)phenoxy-2,3,4,5,6-d5-phosphinyl]-L-Alanine-d5 1-Methylethyl Ester
CAS 1874196-88-9
Molekylær formel C18H16D5N2O7P
Mængde 25 mg
Synonym N-[(S)-(4-Nitrophenoxy)Phenoxy-2,3,4,5,6-D5-Phosphinyl]-L-Alanine-D5 1-Methylethyl Ester
InChI formel InChI=1S/C18H21N2O7P/c1-13(2)25-18(21)14(3)19-28(24,26-16-7-5-4-6-8-16)27-17-11-9-15(10-12-17)20(22)23/h4-14H,1-3H3,(H,19,24)/t14-,28?/m0/s1/i4D,5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(OP(=O)(N[C@@H](C)C(=O)OC(C)C)Oc2ccc(cc2)[N+](=O)[O-])c([2H])c1[2H]
IUPAC navn propan-2-yl (2S)-2-[[(4-nitrophenoxy)-(2,3,4,5,6-pentadeuteriophenoxy)phosphoryl]amino]propanoate
Molekylvægt (g/mol) 413.37
Formel vægt 413.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.