missing translation for 'onlineSavingsMsg'
Learn More

N-[S-(2-Pyridylthio)cysteaminyl]ethylenediamine-N,N,N’,N’-tetraacetic Acid, Monoamide, Technical grade 90%, TRC

Product Code. 30428513
Change view
Click to view available options
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30428513 10 mg
30421212 100 mg
30410000 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428513 Supplier Toronto Research Chemicals Supplier No. TRCP99625010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143541-95-1
Molekylær formel C17 H24 N4 O7 S2
Molekylvægt (g/mol) 460.53
Synonym N-[S-(2-Pyridylthio)Cysteaminyl]Ethylenediamine-N,N,N',N'-Tetraacetic Acid, Monoamide
IUPAC navn 2-[2-[bis(carboxymethyl)amino]ethyl-[2-oxo-2-[2-(pyridin-2-yldisulfanyl)ethylamino]ethyl]amino]acetic acid
SMIL OC(=O)CN(CCN(CC(=O)O)CC(=O)NCCSSc1ccccn1)CC(=O)O

Specifications

Kemisk navn eller materiale N-[S-(2-Pyridylthio)cysteaminyl]ethylenediamine-N,N,N',N'-tetraacetic acid monoamide
Mængde 10 mg
InChI formel InChI=1S/C17H24N4O7S2/c22-13(18-5-8-29-30-14-3-1-2-4-19-14)9-20(10-15(23)24)6-7-21(11-16(25)26)12-17(27)28/h1-4H,5-12H2,(H,18,22)(H,23,24)(H,25,26)(H,27,28)
Formel vægt 460.1086
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.