missing translation for 'onlineSavingsMsg'
Learn More

N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide, 98%, Alfa Aesar™

Brand:  Alfa Aesar H61607.03

Additional Details : CAS Number : 474654-18-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488158

  • 1327.52 DKK / 1g


Estimated Shipment
26-10-2020
Add to basket

Specifications

Chemical Identifiers

474654-18-7
245.71
n-r-phenyl 4-chlorophenyl methyl formamide, n-r-4-chlorophenyl phenylmethyl formamide, n-r-4-chlorophenyl phenyl methyl formamide
C14H12ClNO
HDGOSEHHSVUCBO-UHFFFAOYNA-N

Specifications

N-[(R)-alpha-(4-Chlorophenyl)benzyl]formamide
474654-18-7
n-r-phenyl 4-chlorophenyl methyl formamide, n-r-4-chlorophenyl phenylmethyl formamide, n-r-4-chlorophenyl phenyl methyl formamide
245.71
Solid
1g
C14H12ClNO
HDGOSEHHSVUCBO-UHFFFAOYNA-N
245.7
98%
Videos
SDS Documents