Få mere at vide
n-Propyltriphenylphosphonium-d5 Bromide, TRC
Artikelnummer. 30427286
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30427286 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427286 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8383921G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Propyltriphenylphosphonium-d5 Bromide
CAS 1703757-22-5
Molekylær formel C21 D5 H17 P . Br
Mængde 1 g
Synonym Triphenylpropylphosphonium-d5 Bromide, Bromo(propyl)triphenylphosphorane-d5, NSC 50539-d5, Propyltriphenylphosphonium-d5 Bromide
InChI formel InChI=1S/C21H22P.BrH/c1-2-18-22(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-17H,2,18H2,1H3;1H/q+1;/p-1/i1D3,2D2;
SMIL [Br-].[2H]C([2H])([2H])C([2H])([2H])C[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC navn 2,2,3,3,3-pentadeuteriopropyl(triphenyl)phosphanium;bromide
Molekylvægt (g/mol) 390.3079
Formel vægt 389.0956