Få mere at vide
n-Propyl-d7-amine HCl, CDN
Artikelnummer. 30316774
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316774 0.25 g -
30310988 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316774 Leverandør CDN Leverandørnr. CDND25450.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propylamine-D7 (propyl-D7) Hydrochloride
CAS 344298-88-0
Molekylær formel C3 D7 H2 N . Cl H
Mængde 0.25 g
Synonym 1-Propan-1,1,2,2,3,3,3-d7-amine, hydrochloride (9 CI), Propyl-1,1,2,2,3,3,3-d7-amine hydrochloride, n-Propyl-d7-amine hydrochloride, 1-Propylamine-d7 hydrochloride, Propylamine-D7 (propyl-D7) hydrochloride
InChI formel InChI=1 S/C3H9N.ClH/c1-2-3-4;/h2-4H2,1H3;1 H/i1D3,2D2,3D2;
SMIL Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N
IUPAC navn 1,1,2,2,3,3,3-heptadeuteriopropan-1-amine;hydrochloride
Molekylvægt (g/mol) 102.614
Formel vægt 102.094 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity