missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl acrylate, 95%, stab. with 0.1% 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
n-propyl acrylate is used in coatings, textile and leather finishes, paper coatings, for adhesives, for acrylic fibers. It is used as a monomer for emulsion polymers and solution polymers in latex paints.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Density | 0.92 |
Boiling Point | 43°C to 44°C (40 mmHg) |
Flash Point | 24°C (75°F) |
Refractive Index | 1.4123 |
Quantity | 10 g |
UN Number | UN3272 |
Beilstein | 1744954 |
Solubility Information | Immiscible with water. |
Formula Weight | 114.15 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'