Få mere at vide
n-Propyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Artikelnummer. 30328724
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328724 0.1 g
30321395 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328724 Leverandør CDN Leverandørnr. CDND71140.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxybenzoic-2,3,5,6-D4 Acid Propyl Ester
CAS 1219802-67-1
Molekylær formel C10 H12 O3
Mængde 0.1 g
Synonym 4-Hydroxybenzoic acid-propyl ester D4
InChI formel InChI=1 S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11 H,2,7H2,1H3/i3D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c(C(=O)OCCC)c([2 H])c([2 H])c1O
IUPAC navn propyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 184.2251
Formel vægt 184.1038 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity