Få mere at vide
n-Propyl-3,3,3-d3-amine, CDN
Artikelnummer. 30310389
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310389 0.1 g
30327804 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310389 Leverandør CDN Leverandørnr. CDND57000.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Propylamine-d3
CAS 24300-22-9
Molekylær formel C3 D3 H6 N
Mængde 0.1 g
Synonym 1-Propan-3,3,3-d3-amine (9 CI), Propyl-3,3,3-d3-amine (7 CI, 8 CI), n-Propyl-3,3,3-d3-amine, 1-Propylamine-d3, Propylamine-D3 (propyl-3,3,3-D3)
InChI formel InChI=1 S/C3H9N/c1-2-3-4/h2-4H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCN
IUPAC navn 3,3,3-trideuteriopropan-1-amine
Molekylvægt (g/mol) 62.129
Formel vægt 62.092 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity