missing translation for 'onlineSavingsMsg'
Få mere at vide

n-Propyl-2-d1 Alcohol, CDN

Artikelnummer. 30316874
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316874 0.25 g
30318522 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316874 Leverandør CDN Leverandørnr. CDND56520.25G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale 1-Propanol-2-D
CAS 89603-83-8
Molekylær formel C3 D H7 O
Mængde 0.25 g
Synonym 1-Propan-2-d-ol (9 CI), Propyl-2-d-alcohol (7 CI), 2-Monodeuteropropanol, 2-Monodeuteropropan-1-ol, 1-Propanol-2-D
InChI formel InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i2D
SMIL [2 H]C(C)CO
IUPAC navn 2-deuteriopropan-1-ol
Molekylvægt (g/mol) 61.101
Formel vægt 61.064 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.