Få mere at vide
n-Propyl-2,2,3,3,3-d5-amine, CDN
Artikelnummer. 30325767
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325767 0.25 g
30322539 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325767 Leverandør CDN Leverandørnr. CDND74620.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propylamine-D5 (propyl-2,2,3,3,3-D5)
CAS 1398066-17-5
Molekylær formel C3 D5 H4 N
Mængde 0.25 g
Synonym 1-Propan-2,2,3,3,3-d5-amine, Propyl-2,2,3,3,3-d5-amine, n-Propyl-d5-amine, 1-Propylamine-2,2,3,3,3-d5, Propylamine-D5 (propyl-2,2,3,3,3-D5)
InChI formel InChI=1 S/C3H9N/c1-2-3-4/h2-4H2,1H3/i1D3,2D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])CN
IUPAC navn 2,2,3,3,3-pentadeuteriopropan-1-amine
Molekylvægt (g/mol) 64.141
Formel vægt 64.105 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity