Få mere at vide
n-Propyl-1,1,2,2-d4 Alcohol, CDN
Artikelnummer. 30324382
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324382 0.25 g -
30319197 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324382 Leverandør CDN Leverandørnr. CDND58240.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Propanol-1,1,2,2-D4
CAS 70907-80-1
Molekylær formel C3 D4 H4 O
Mængde 0.25 g
Synonym 1-Propan-1,1,2,2-d4-ol (9 CI), 1,1,2,2-Tetradeuteropropanol, 1,1,2,2-Tetradeuteropropan-1-ol, Propyl-1,1,2,2-d4 alcohol, 1-Propanol-1,1,2,2-D4
InChI formel InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i2D2,3D2
SMIL [2 H]C([2 H])(C)C([2 H])([2 H])O
IUPAC navn 1,1,2,2-tetradeuteriopropan-1-ol
Molekylvægt (g/mol) 64.12
Formel vægt 64.083 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity