Learn More
N-Propionyl-L-cysteine, Mikromol™
Product Code. 30050260
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050260 100 mg 100mg
30049195 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050260 Supplier Mikromol Supplier No. MM0024.06
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Propionyl-L-cysteine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Propionyl-L-cysteine
CAS 2885-79-2
Analyte or Component Names N-Propionyl-L-cysteine
Shipping Condition Room Temperature
Molecular Formula C6 H11 N O3 S
Certifications/Compliance ISO 9001
Synonym N-Propionyl-L-cysteine
InChI Formula InChI=1S/C6H11NO3S/c1-2-5(8)7-4(3-11)6(9)10/h4,11H,2-3H2,1H3,(H,7,8)(H,9,10)/t4-/m0/s1
SMILES CCC(=O)N[C@@H](CS)C(=O)O
IUPAC Name (2R)-2-(propanoylamino)-3-sulfanylpropanoic acid
Molecular Weight (g/mol) 177.22
Quantity 100 mg
Formula Weight 177.046 g/mol