missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Phthaloylglycine, 98+%

Product Code. 11448456
Change view
Click to view available options
Mængde:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
11448456 50 g 50g
11458456 250 g 250g
11438456 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448456 Supplier Thermo Scientific Alfa Aesar Supplier No. A15920.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4702-13-0 | C10H7NO4 | 205.169 g/mol

Denne Thermo Scientific Chemicals mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific Chemicals .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4702-13-0
Molekylær formel C10H7NO4
Molekylvægt (g/mol) 205.169
MDL nummer MFCD00005900
InChI nøgle WQINSVOOIJDOLJ-UHFFFAOYSA-N
Synonym n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid
PubChem CID 20825
IUPAC navn 2-(1,3-dioxoisoindol-2-yl)eddikesyre
SMIL C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O

Specifications

Kemisk navn eller materiale N-Phthaloylglycine
Smeltepunkt 194°C to 198°C
Beilstein 184174
Mængde 50 g
Formel vægt 205.17
Procent renhed ≥98%

RUO– Kun brug til forskning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.