Få mere at vide
N-Phenylacetylglycine, TRC
Artikelnummer. 30132639
Skift visning
Mængde:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132639 100 mg
30137445 500 mg
30009780 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132639 Leverandør Toronto Research Chemicals Leverandørnr. TRCP306550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500-98-1
Molekylær formel C10 H11 N O3
Molekylvægt (g/mol) 193.2
Synonym Glycine, N-(2-phenylacetyl)-, Glycine, N-(phenylacetyl)- (8CI,9CI), Phenaceturic acid (6CI), N-(2-Phenylacetyl)glycine, (2-Phenylacetyl)glycine, 2-(2-Phenylacetamido)acetic acid, 2-[(2-Phenylacetyl)amino]acetic acid, N-(Phenylacetyl)glycine, N-Phenacetylglycine, NSC 408424, NSC 92778, Phenacetylglycine, Phenylacetylglycine
IUPAC navn 2-[(2-phenylacetyl)amino]acetic acid
SMIL OC(=O)CNC(=O)Cc1ccccc1
Tekniske data
Kemisk navn eller materiale N-Phenylacetylglycine
InChI formel InChI=1S/C10H11NO3/c12-9(11-7-10(13)14)6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)(H,13,14)
Mængde 100 mg
Formel vægt 193.0739