Få mere at vide
N-(Phenylacetyl-d5)glycine, TRC
Artikelnummer. 30180223
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180223 1 mg
30180100 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180223 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3065521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189920-31-7
Molekylær formel C10 D5 H6 N O3
Molekylvægt (g/mol) 198.23
Synonym 2-[[2-(2,3,4,5,6-Pentadeuteriophenyl)acetyl]amino]acetic acid, 2-(2-Phenylacetamido)acetic acid D5 (phenyl-D5), Phenaceturic acid-d5, N-(2-Phenyl-d5-acetyl)glycine, (2-Phenylacetyl)glycine-d5, 2-(2-Phenylacetamido)acetic acid-d5, 2-[[2-(Phenyl-d5)acetyl]amino]acetic acid, N-(Phenyl-d5-acetyl)glycine, N-Phenacetylglycine-d5, Phenacetylglycine-d5, Phenylacetylglycine-d5
IUPAC navn 2-[[2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]acetic acid
SMIL [2H]c1c([2H])c([2H])c(CC(=O)NCC(=O)O)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale N-(Phenyl-d5-acetyl)glycine
InChI formel InChI=1S/C10H11NO3/c12-9(11-7-10(13)14)6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)(H,13,14)/i1D,2D,3D,4D,5D
Mængde 1 mg
Formel vægt 198.105