Learn More
N-Phenyl-3-(trifluoromethyl)aniline, Mikromol™
Product Code. 30051343
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30051343 100 mg 100mg
1 options
Product Code. 30051343 Supplier Mikromol Supplier No. MM0105.03
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Phenyl-3-(trifluoromethyl)aniline
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Phenyl-3-(trifluoromethyl)aniline
CAS 101-23-5
Analyte or Component Names N-Phenyl-3-(trifluoromethyl)aniline
Shipping Condition Room Temperature
Molecular Formula C13 H10 F3 N
Certifications/Compliance ISO 17025
Synonym N-Phenyl-3-(trifluoromethyl)aniline, Etofenamate Imp. C (EP), Etofenamate Impurity C
InChI Formula InChI=1S/C13H10F3N/c14-13(15,16)10-5-4-8-12(9-10)17-11-6-2-1-3-7-11/h1-9,17H
SMILES FC(F)(F)c1cccc(Nc2ccccc2)c1
IUPAC Name N-phenyl-3-(trifluoromethyl)aniline
Molecular Weight (g/mol) 237.22
Quantity 100 mg
Formula Weight 237.0765 g/mol