missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Phenyl-3-(trifluoromethyl)aniline, 95%, BLD Pharm™

Artikelnummer. 31028602
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
31028602 1 g
31021462 5 g
31023221 100 mg
31024900 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31028602 Leverandør BLD Pharm Leverandørnr. BD1455541g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Specifications

Kemisk navn eller materiale N-Phenyl-3-(trifluoromethyl)aniline
CAS 101-23-5
Molekylær formel C13H10F3N
MDL nummer MFCD00017998
Mængde 1 g
SMIL FC(C1=CC(NC2=CC=CC=C2)=CC=C1)(F)F
Molekylvægt (g/mol) 237.22
Procent renhed 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.