missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Phenyl 2-Bromo-5-Fluorobenzamide, TRC

Artikelnummer. 30418425
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30418425 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30418425 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP400985500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Phenyl 2-Bromo-5-Fluorobenzamide
CAS 949443-48-5
Molekylær formel C13 H9 Br F N O
Mængde 500 mg
InChI formel InChI=1S/C13H9BrFNO/c14-12-7-6-9(15)8-11(12)13(17)16-10-4-2-1-3-5-10/h1-8H,(H,16,17)
SMIL Fc1ccc(Br)c(c1)C(=O)Nc2ccccc2
IUPAC navn 2-bromo-5-fluoro-N-phenylbenzamide
Molekylvægt (g/mol) 294.119
Formel vægt 292.985
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.