Få mere at vide
N-Phenoxyisopropyl-N-benzylaziridinium Perchlorate, TRC
Artikelnummer. 30423402
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423402 10 mg
30413492 50 mg
30417355 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423402 Leverandør Toronto Research Chemicals Leverandørnr. TRCP39972110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57619-17-7
Molekylær formel C18 H22 N O . Cl O4
Molekylvægt (g/mol) 367.82
Synonym 1-Benzyl-1-(1-phenoxypropan-2-yl)aziridin-1-ium Perchlorate
IUPAC navn 1-benzyl-1-(1-methyl-2-phenoxy-ethyl)aziridin-1-ium;perchlorate
SMIL CC(COc1ccccc1)[N+]2(Cc3ccccc3)CC2.[O-]Cl(=O)(=O)=O
Tekniske data
Kemisk navn eller materiale N-Phenoxyisopropyl-N-benzylaziridinium perchlorate
Mængde 10 mg
InChI formel InChI=1S/C18H22NO.ClHO4/c1-16(15-20-18-10-6-3-7-11-18)19(12-13-19)14-17-8-4-2-5-9-17;2-1(3,4)5/h2-11,16H,12-15H2,1H3;(H,2,3,4,5)/q+1;/p-1
Formel vægt 367.1186