Få mere at vide
N-Pentylindole-d11, TRC
Artikelnummer. 30418938
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30418938 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418938 Leverandør Toronto Research Chemicals Leverandørnr. TRCP28020250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Pentylindole-d11
CAS 1346603-11-9
Molekylær formel C13H6D11N
Mængde 50 mg
Synonym 1-Pentylindole-d11, 1-Pentyl-1H-indole-d11
InChI formel InChI=1S/C13H17N/c1-2-3-6-10-14-11-9-12-7-4-5-8-13(12)14/h4-5,7-9,11H,2-3,6,10H2,1H3/i1D3,2D2,3D2,6D2,10D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])n1ccc2ccccc12
IUPAC navn 1-(1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl)indole
Molekylvægt (g/mol) 198.35
Formel vægt 198.2051