Få mere at vide
n-Pentylbenzene-2,3,4,5,6-d5, TRC
Artikelnummer. 30412672
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30412672 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412672 Leverandør Toronto Research Chemicals Leverandørnr. TRCP279801100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Pentylbenzene-2,3,4,5,6-d5
CAS 68639-73-6
Molekylær formel C11 D5 H11
Mængde 100 mg
Synonym Benzene-d5','pentyl- (9CI), Pentylbenzene-d5, Pentylbenzene-2,3,4,5,6-D5, 1-(Phenyl-d5)pentane, Amylbenzene-d5, n-Pentylbenzene-2,3,4,5,6-d5
InChI formel InChI=1S/C11H16/c1-2-3-5-8-11-9-6-4-7-10-11/h4,6-7,9-10H,2-3,5,8H2,1H3/i4D,6D,7D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(CCCCC)c([2H])c1[2H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-pentylbenzene
Molekylvægt (g/mol) 153.2756
Formel vægt 153.1566