Få mere at vide
n-Pentylbenzene-2,3,4,5,6-d5, CDN
Artikelnummer. 30319302
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319302 0.5 g
30311993 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319302 Leverandør CDN Leverandørnr. CDND66530.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Pentylbenzene-2,3,4,5,6-d5
CAS 68639-73-6
Molekylær formel C11 D5 H11
Mængde 0.5 g
Synonym Benzene-d5, pentyl- (9 CI), Pentylbenzene-d5, Pentylbenzene-2,3,4,5,6-D5, 1-(Phenyl-d5)pentane, Amylbenzene-d5, n-Pentylbenzene-2,3,4,5,6-d5
InChI formel InChI=1 S/C11H16/c1-2-3-5-8-11-9-6-4-7-10-11/h4,6-7,9-10 H,2-3,5,8H2,1H3/i4D,6 D,7 D,9 D,10 D
SMIL [2 H]c1c([2 H])c([2 H])c(CCCCC)c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-pentylbenzene
Molekylvægt (g/mol) 153.2756
Formel vægt 153.1566 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity