Få mere at vide
n-Pentyl-d11 Chloroformate, CDN
Artikelnummer. 30316898
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316898 0.25 g
30310069 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316898 Leverandør CDN Leverandørnr. CDND75140.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentyl Chloroformate D11
CAS 1398065-67-2
Molekylær formel C6 D11 Cl O2
Mængde 0.25 g
Synonym Amyl-d11 chloroformate, Chloroformic acid amyl ester-d11, Chloroformic acid pentyl-d11 ester, n-Pentyl-d11 chloroformate, Pentyl chloroformate D11, Undecadeuteriopentyloxycarbonyl chloride, Carbonochloridic acid pentyl-d11 ester
InChI formel InChI=1 S/C6H11ClO2/c1-2-3-4-5-9-6(7)8/h2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OC(=O)Cl
IUPAC navn 1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl carbonochloridate
Molekylvægt (g/mol) 161.671
Formel vægt 161.114 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity