Få mere at vide
n-Pentyl Alcohol-OD, CDN
Artikelnummer. 30318066
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30318066 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318066 Leverandør CDN Leverandørnr. CDND72425G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Pentanol-OD
CAS 14848-79-4
Molekylær formel C5 D H11 O
Mængde 5 g
Synonym 1-Pentanol-d (9 CI, ACI), Pentyl alcohol-d (8 CI), n-Pentyl alcohol-OD, 1-Pentanol-OD
InChI formel InChI=1 S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i6D
SMIL [2 H]OCCCCC
IUPAC navn 1-deuteriooxypentane
Molekylvægt (g/mol) 89.154
Formel vægt 89.095 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity