Få mere at vide
n-Pentyl Alcohol-d12, CDN
Artikelnummer. 30324509
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324509 0.5 g
30311635 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324509 Leverandør CDN Leverandørnr. CDND58500.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Pentanol-D12
CAS 158778-85-9
Molekylær formel C5 D12 O
Mængde 0.5 g
Synonym 1-Pentan-1,1,2,2,3,3,4,4,5,5,5-d11-ol-d (9 CI), 1-Pentanol-d12, 1-Pentan-d11-ol-d, n-Pentyl-d11 alcohol-OD, n-Pentyl alcohol-d12
InChI formel InChI=1 S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2,6 D
SMIL [2 H]OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5-undecadeuterio-5-deuteriooxypentane
Molekylvægt (g/mol) 100.2221
Formel vægt 100.1641 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity