Få mere at vide
n-Pentyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Artikelnummer. 30314267
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314267 0.05 g -
30320162 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314267 Leverandør CDN Leverandørnr. CDND72380.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentyl 4-Hydroxybenzoate D4 (phenyl-2,3,5,6-D4)
CAS 1219798-66-9
Molekylær formel C12 D4 H12 O3
Mængde 0.05 g
Synonym Pentyl 4-hydroxy-2,3,5,6-tetradeuteriobenzoate, Pentyl 4-hydroxybenzoate D4 (phenyl-2,3,5,6-D4), Pentyl 4-hydroxybenzoate-2,3,5,6-d4, Pentyl p-hydroxybenzoate-d4, Pentyl paraben-d4, 4-Hydroxybenzoic-2,3,5,6-d4 acid pentyl ester
InChI formel InChI=1 S/C12H16O3/c1-2-3-4-9-15-12(14)10-5-7-11(13)8-6-10/h5-8,13 H,2-4,9H2,1H3/i5D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c(C(=O)OCCCCC)c([2 H])c([2 H])c1O
IUPAC navn pentyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 212.278
Formel vægt 212.135 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity