missing translation for 'onlineSavingsMsg'
Learn More

N-Pentane, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/1020/PB17

Additional Details : CAS Number : 109-66-0 Weight : 1.56500kg Transport : UN number : 1265 Chem class : 3 Pack group : II


Packaging Plastic Coated Glass bottle
Quantity 2.5L

 View more versions of this product

Product Code. 10030662

  • 680.37 DKK / 2.50 L

Estimated Shipment: 27-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

N-Pentane, Extra Pure, SLR

Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
CHEBI:37830
CCCCC
C5H12
MFCD00009498
8003
pentane

Specifications

109-66-0
MFCD00009498
CCCCC
72.15
CHEBI:37830
Liquid
573mbar at 20°C
2.5
36°C
2.5L
C5H12
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15g/mol
Plastic Coated Glass bottle
0.25 mPaS at 20°C
-130°C
Undesignated
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • n-Pentane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

SDS
Documents
Certificates