Få mere at vide
N-Pentane-d12, TRC
Artikelnummer. 30425657
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30425657 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425657 Leverandør Toronto Research Chemicals Leverandørnr. TRCP274055500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Pentane-d12
CAS 2031-90-5
Molekylær formel C5 D12
Mængde 500 mg
Synonym Pentane-1,1,1,2,2,3,3,4,4,5,5,5-d12 (ACI), Pentane-d12 (7CI','8CI','9CI), 1,1,1,2,2,3,3,4,4,5,5,5-Dodecadeuteriopentane, n-Pentane-d12, Perdeuteriopentane, n-Pentane-D12
InChI formel InChI=1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3/i1D3,2D3,3D2,4D2,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,5-dodecadeuteriopentane
Molekylvægt (g/mol) 84.22
Formel vægt 84.1692