missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Palmitoyl-O-phosphocholine Serine-D9, TRC

Artikelnummer. 30428374
missing translation for 'orderingAttributeHoverText'
Mængde:
1 mg
5 mg
10 mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30428374

missing translation for 'mfr': Toronto Research Chemicals TRCP3479211MG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Palmitoyl-O-phosphocholine Serine-D9
Molekylær formel C24 D9 H40 N2 O7 P
Mængde 1 mg
Synonym (S)-2-Carboxy-2-palmitamidoethyl (2-(Tris(methyl-d3)ammonio)ethyl)phosphate
InChI formel InChI=1S/C24H49N2O7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(27)25-22(24(28)29)21-33-34(30,31)32-20-19-26(2,3)4/h22H,5-21H2,1-4H3,(H2-,25,27,28,29,30,31)/t22-/m0/s1/i2D3,3D3,4D3
SMIL [2H]C([2H])([2H])[N+](CCOP(=O)([O-])OC[C@H](NC(=O)CCCCCCCCCCCCCCC)C(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn [(2S)-2-carboxy-2-(hexadecanoylamino)ethyl] 2-[tris(trideuteriomethyl)azaniumyl]ethyl phosphate
Molekylvægt (g/mol) 517.684
Formel vægt 517.384
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.