Få mere at vide
N-p-Coumaroyltyrosine (~98%), TRC
Artikelnummer. 30365852
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365852 50 mg
30366316 10 mg
30349674 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365852 Leverandør Toronto Research Chemicals Leverandørnr. TRCC89998050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 77201-66-2
Molekylær formel C18H17NO5
Molekylvægt (g/mol) 327.33
Synonym N-[(2E)-3-(4-Hydroxyphenyl)-1-oxo-2-propen-1-yl]-L-tyrosine, N-[(2E)-3-(4-hydroxyphenyl)-1-oxo-2-propenyl]-L-tyrosine, (E)-N-[3-(4-Hydroxyphenyl)-1-oxo-2-propenyl]-L-tyrosine
IUPAC navn (2S)-3-(4-hydroxyphenyl)-2-[[(E)-3-(4-hydroxyphenyl)prop-2-enoyl]amino]propanoic acid
SMIL OC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)\C=C\c2ccc(O)cc2
Tekniske data
Kemisk navn eller materiale N-p-Coumaroyltyrosine
Mængde 50 mg
InChI formel InChI=1S/C18H17NO5/c20-14-6-1-12(2-7-14)5-10-17(22)19-16(18(23)24)11-13-3-8-15(21)9-4-13/h1-10,16,20-21H,11H2,(H,19,22)(H,23,24)/b10-5+/t16-/m0/s1
Formel vægt 327.1107