missing translation for 'onlineSavingsMsg'
Learn More

N-[p-(Acetylmercuric)phenyl]maleimide, TRC

Product Code. 30312156
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30312156 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312156 Supplier Toronto Research Chemicals Supplier No. TRCA18628050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-[p-(Acetylmercuric)phenyl]maleimide
CAS 117259-61-7
Molekylær formel C12H9HgNO4
Mængde 50 mg
Synonym (Acetato-O)[4-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)phenyl]mercury, 1-Phenyl-1H-Pyrrole-2,5-dione Mercury Complex
InChI formel InChI=1S/C10H6NO2.C2H4O2.Hg/c12-9-6-7-10(13)11(9)8-4-2-1-3-5-8;1-2(3)4;/h2-7H;1H3,(H,3,4);/q;;+1/p-1
SMIL CC(=O)O[Hg]c1ccc(cc1)N2C(=O)C=CC2=O
IUPAC navn acetyloxy-[4-(2,5-dioxopyrrol-1-yl)phenyl]mercury
Molekylvægt (g/mol) 431.79
Formel vægt 433.0238
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.