Få mere at vide
N-Oleoylethanolamide-d4, TRC
Artikelnummer. 30415880
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415880 5 mg
30427616 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415880 Leverandør Toronto Research Chemicals Leverandørnr. TRCO5272825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Oleoyl Ethanolamide-d4
CAS 946524-36-3
Molekylær formel C20 D4 H35 N O2
Mængde 5 mg
Synonym (9Z)-N-(2-Hydroxyethyl-1,1,2,2-d4)-9-octadecenamide, (Z)-N-(2-Hhydroxyethyl-d4)-9-octadecenamide, N-(2-Hydroxyethyl-d4)-oleamide, AM 3101, N-(2-Hydroxyethyl-d4)-9-Z-octadecenamide, N-(2-Hydroxyethyl-d4)oleamide, N-Oleoyl-2-aminoethanol-d4, N-Oleoylethanol-d4-amide, N-Oleoylethanol-d4-amine, Oleamide MEA-d4, Oleic Acid Ethanol-d4-amide, Oleic Acid Monoethanol-d4-amide, Oleoylethanol-d4-amide
InChI formel InChI=1S/C20H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h9-10,22H,2-8,11-19H2,1H3,(H,21,23)/b10-9-/i18D2,19D2
SMIL [2H]C([2H])(O)C([2H])([2H])NC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC navn (Z)-N-(1,1,2,2-tetradeuterio-2-hydroxyethyl)octadec-9-enamide
Molekylvægt (g/mol) 329.554
Formel vægt 329.323