Få mere at vide
n-Octyl-oligo-oxyethylene, TRC
Artikelnummer. 30428607
Skift visning
Mængde:
250 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428607 250 mg
30423450 2.5 g
30419148 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428607 Leverandør Toronto Research Chemicals Leverandørnr. TRCO293273250MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27252-75-1
Molekylær formel (C2H4O)nC8H18O
Molekylvægt (g/mol) 174.28
Synonym Aduxol Hex 05B, Berol EP 35, Dehydol 04, Dehydol 04DEO, Ethoxylated Octyl Alcohol, Novel II 8-7, Pionin D 1002, Poly(oxyethylene) Monooctyl Ether, Poly(oxyethylene) Octyl Ether, Polyethoxylated Octanol, Polyethylene Glycol Mono-n-octyl Ether, Polyethylene Glycol Monooctyl Ether, Polyethylene Glycol Octyl Ether, Polyethylene Glycol Octyl Monoether, Polyoxyethylene Mono(n-octyl) Ether, THM Schkopau W-OF, Texofor N, Texofor N 12, Texofor N 4, Trycol 5887, Trycol ST 8049, n-Octyl Polyethylene Glycol Ether, n-Octylpolyoxyethylene, alpha-Octyl-ω-hydroxy-poly(oxyethylene), alpha-Octyl-ω-hydroxypoly(oxy-1,2-ethanediyl)
IUPAC navn 2-octoxyethanol
SMIL CCCCCCCCOCCO
Tekniske data
Kemisk navn eller materiale n-Octyl-oligo-oxyethylene
Mængde 250 mg
InChI formel InChI=1S/C10H22O2/c1-2-3-4-5-6-7-9-12-10-8-11/h11H,2-10H2,1H3
Formel vægt 174.162