Få mere at vide
n-Octadecyl-d37 Alcohol, TRC
Artikelnummer. 30419082
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30419082 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419082 Leverandør Toronto Research Chemicals Leverandørnr. TRCO2354815MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Octadecanol-D37
CAS 204259-62-1
Molekylær formel C18HD37O
Mængde 5 mg
Synonym 1-Octadecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,18-d37-ol (9CI), Stearyl-d37 alcohol, 1-Octadecanol-d37, 1-Octadecanol-D37, n-Octadecyl-d37 alcohol
InChI formel InChI=1S/C18H38O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h19H,2-18H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2
SMIL OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn octadecan-d37-1-ol
Molekylvægt (g/mol) 307.73
Formel vægt 307.52