Få mere at vide
N,O-Ditrityl Losartan, TRC
Artikelnummer. 30373511
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30373511 250 mg
30370681 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30373511 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494320250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1796930-34-1
Molekylær formel C60 H51 Cl N6 O
Molekylvægt (g/mol) 907.54
Synonym 2H-Tetrazole, 5-[4'-[[2-butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-, 5-[4'-[[2-Butyl-4-chloro-5-[[(triphenylmethyl)oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-2-(triphenylmethyl)tetrazole, 5-[4'-[[2-Butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-2H-tetrazole, 2H-Tetrazole, 5-[4'-[[2-butyl-4-chloro-5-[(triphenylmethoxy)methyl]-1H-imidazol-1-yl]methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-, 5-[4'-[[2-Butyl-4-chloro-5-[[(triphenylmethyl)oxy]methyl]-1H-imidazol-1-yl]methyl]biphenyl-2-yl]-2-(triphenylmethyl)tetrazole
IUPAC navn 5-[2-[4-[[2-butyl-4-chloro-5-(trityloxymethyl)imidazol-1-yl]methyl]phenyl]phenyl]-2-trityltetrazole
SMIL CCCCc1nc(Cl)c(COC(c2ccccc2)(c3ccccc3)c4ccccc4)n1Cc5ccc(cc5)c6ccccc6c7nnn(n7)C(c8ccccc8)(c9ccccc9)c%10ccccc%10
Tekniske data
Kemisk navn eller materiale N,O-Ditrityl Losartan
Mængde 250 mg
InChI formel InChI=1S/C60H51ClN6O/c1-2-3-38-56-62-57(61)55(44-68-60(50-30-16-7-17-31-50,51-32-18-8-19-33-51)52-34-20-9-21-35-52)66(56)43-45-39-41-46(42-40-45)53-36-22-23-37-54(53)58-63-65-67(64-58)59(47-24-10-4-11-25-47,48-26-12-5-13-27-48)49-28-14-6-15-29-49/h4-37,39-42H,2-3,38,43-44H2,1H3
Formel vægt 906.3813